missing translation for 'onlineSavingsMsg'
Learn More

6-Chloropiperonyl alcohol, 98%

Product Code. 11302316
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11302316 25 g 25g
11392306 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11302316 Supplier Thermo Scientific Alfa Aesar Supplier No. L05972.14
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 2591-25-5 | C8H7ClO3 | 186.591 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2591-25-5
Molecular Formula C8H7ClO3
Molecular Weight (g/mol) 186.591
MDL Number MFCD00051727
InChI Key ADHYYNHCXPPVHQ-UHFFFAOYSA-N
Synonym 6-chloropiperonyl alcohol, 6-chloro-2h-1,3-benzodioxol-5-yl methanol, 6-chlorobenzo d 1,3 dioxol-5-yl methanol, 6-chloro-1,3-benzodioxol-5-yl methanol, 2-chlor-4,5-methylendioxy-benzylalkohol, 6-chloro-3,4-methylenedioxybenzyl alcohol, 6-chloro-2h-benzo d 1,3-dioxolen-5-yl methan-1-ol
PubChem CID 7015319
IUPAC Name (6-chloro-1,3-benzodioxol-5-yl)methanol
SMILES C1OC2=C(O1)C=C(C(=C2)CO)Cl

Specifications

Melting Point 68°C to 72°C
Quantity 25 g
Beilstein 150801
Formula Weight 186.6
Percent Purity 98%
Chemical Name or Material 6-Chloropiperonyl alcohol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.