missing translation for 'onlineSavingsMsg'
Learn More
Learn More
6-chlorooxindole, 98%, ACROS Organics™
£28.23 - £101.00
Chemical Identifiers
CAS | 56341-37-8 |
---|---|
Molecular Formula | C8H6ClNO |
Molecular Weight (g/mol) | 167.59 |
MDL Number | MFCD00209962 |
InChI Key | CENVPIZOTHULGJ-UHFFFAOYSA-N |
Synonym | 6-chlorooxindole, 6-chloroindolin-2-one, 6-chloro-2-oxindole, 6-chloro-2-indolinone, 6-chloro-1,3-dihydro-2h-indol-2-one, 6-chloro 2-oxindole, 2h-indol-2-one, 6-chloro-1,3-dihydro, 6-chloro-1,3-dihydro-indol-2-one, 6-chloroindole-2 3h-one, 6-chloro-2,3-dihydro-1h-indol-2-one |
PubChem CID | 736344 |
IUPAC Name | 6-chloro-1,3-dihydroindol-2-one |
SMILES | C1C2=C(C=C(C=C2)Cl)NC1=O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10612445
|
Acros Organics
399440050 |
5g | Glass bottle |
£28.23
5g |
Estimated Shipment 16-02-2021 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
10041644
|
Acros Organics
399440250 |
25g | Glass bottle |
£101.00
25g |
In stock, ready to ship: 2 | Please sign in to purchase this item. Need a web account? Register with us today! | |||
Chemical Identifiers
56341-37-8 | |
167.59 | |
CENVPIZOTHULGJ-UHFFFAOYSA-N | |
736344 | |
C1C2=C(C=C(C=C2)Cl)NC1=O |
C8H6ClNO | |
MFCD00209962 | |
6-chlorooxindole, 6-chloroindolin-2-one, 6-chloro-2-oxindole, 6-chloro-2-indolinone, 6-chloro-1,3-dihydro-2h-indol-2-one, 6-chloro 2-oxindole, 2h-indol-2-one, 6-chloro-1,3-dihydro, 6-chloro-1,3-dihydro-indol-2-one, 6-chloroindole-2 3h-one, 6-chloro-2,3-dihydro-1h-indol-2-one | |
6-chloro-1,3-dihydroindol-2-one |
Specifications
6-chlorooxindole | |
195°C to 199°C | |
56341-37-8 | |
100.0 | |
C8H6ClNO | |
6-chlorooxindole, 6-chloroindolin-2-one, 6-chloro-2-oxindole, 6-chloro-2-indolinone, 6-chloro-1,3-dihydro-2h-indol-2-one, 6-chloro 2-oxindole, 2h-indol-2-one, 6-chloro-1,3-dihydro, 6-chloro-1,3-dihydro-indol-2-one, 6-chloroindole-2 3h-one, 6-chloro-2,3-dihydro-1h-indol-2-one | |
C1C2=C(C=C(C=C2)Cl)NC1=O | |
167.59 | |
167.59 |
Glass bottle | |
5g | |
97.5 | |
97.5% min. (HPLC) | |
MFCD00209962 | |
CENVPIZOTHULGJ-UHFFFAOYSA-N | |
6-chloro-1,3-dihydroindol-2-one | |
736344 | |
98% |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- 6-Chlorooxindole
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Respiratory sensitiser Category 1
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H317-May cause an allergic skin reaction.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H302-Harmful if swallowed.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.