missing translation for 'onlineSavingsMsg'
Learn More

6-Chloronicotinoyl chloride, 98%, Thermo Scientific Chemicals

Product Code. 11459483
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 58757-38-3
Molecular Formula C6H3Cl2NO
Molecular Weight (g/mol) 175.996
MDL Number MFCD00051775
InChI Key FMEBIWNKYZUWFV-UHFFFAOYSA-N
Synonym 6-chloronicotinoyl chloride, 6-chloronicotinoylchloride, 2-chloropyridine-5-carbonyl chloride, 3-pyridinecarbonylchloride, 6-chloro, 3-pyridinecarbonyl chloride, 6-chloro, 6-chloro-nicotinoyl chloride, pubchem18665, 6-chloronicotinic chloride, acmc-1ay8e, 6-chloro-nicotinic chloride
PubChem CID 2782165
IUPAC Name 6-chloropyridine-3-carbonyl chloride
SMILES C1=CC(=NC=C1C(=O)Cl)Cl

Specifications

Melting Point 48°C to 52°C
Boiling Point 84°C to 85°C (3 mmHg)
Quantity 5 g
UN Number UN3261
Beilstein 115927
Formula Weight 176
Percent Purity 98%
Chemical Name or Material 6-Chloronicotinoyl chloride

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'