missing translation for 'onlineSavingsMsg'
Learn More

6-Chloronicotinic Acid, TRC

Product Code. 30136770
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30136770 100 g 100g
30134939 25 g 25g
17924215 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30136770 Supplier Toronto Research Chemicals Supplier No. TRCC370930100G
 Restricted Item
Estimated Shipment: 21-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5326-23-8
Molecular Formula C6 H4 Cl N O2
Molecular Weight (g/mol) 157.55
Synonym Nicotinic acid, 6-chloro- (6CI,7CI,8CI), 6-Chloro-3-pyridinecarboxylic acid, 2-Chloro-5-pyridinecarboxylic acid, 3-Carboxy-6-chloropyridine, 6-Chloronicotinic acid, 6-Chloropyridin-3-carboxylic acid, NSC 277
IUPAC Name 6-chloropyridine-3-carboxylic acid
SMILES OC(=O)c1ccc(Cl)nc1

Specifications

Quantity 100 g
InChI Formula InChI=1S/C6H4ClNO2/c7-5-2-1-4(3-8-5)6(9)10/h1-3H,(H,9,10)
Formula Weight 156.9931
Chemical Name or Material 6-Chloronicotinic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.