missing translation for 'onlineSavingsMsg'
Learn More

6-Chloronicotinic acid, 99%

Product Code. 10782971 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10782971 25 g Glass bottle 25g
10225790 5 g Glass bottle 5g
10062050 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10782971 Supplier Thermo Scientific Acros Supplier No. 168690250
 Restricted Item
Estimated Shipment: 20-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 5326-23-8 | C6H4ClNO2 | 157.56 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5326-23-8
Molecular Formula C6H4ClNO2
Molecular Weight (g/mol) 157.56
MDL Number MFCD00006241
InChI Key UAWMVMPAYRWUFX-UHFFFAOYSA-N
Synonym 6-chloronicotinic acid, 6-chloro-3-pyridinecarboxylic acid, 3-pyridinecarboxylic acid, 6-chloro, 6-chloro-nicotinic acid, 6-chloro nicotinic acid, 2-chloro-5-pyridinecarboxylic acid, 2-chloropyridine-5-carboxylic acid, 6-chlornicotins, 6chloronichotinicacid, 2-chloroncotinic acid
PubChem CID 79222
IUPAC Name 6-chloropyridine-3-carboxylic acid
SMILES C1=CC(=NC=C1C(=O)O)Cl

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 186°C to 190°C
Color White to Beige
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (HPLC)
Packaging Glass bottle
Linear Formula Cl(C5H3N)COOH
Quantity 25 g
Beilstein 22, 43
Solubility Information Solubility in water: 2g/L (20°C)
Formula Weight 157.56
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material 6-Chloronicotinic acid, 99%
Show More Show Less
compliance-icons
Product Identifier
  • 6-Chloronicotinic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.