missing translation for 'onlineSavingsMsg'
Learn More

6-Chloronicotinamide, 98%

Product Code. 11455007
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11455007 1 g 1g
11465007 5 g 5g
11475007 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11455007 Supplier Thermo Scientific Alfa Aesar Supplier No. B21922.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 6271-78-9 | C6H5ClN2O | 156.57 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6271-78-9
Molecular Formula C6H5ClN2O
Molecular Weight (g/mol) 156.57
MDL Number MFCD00006242
InChI Key ZIJAZUBWHAZHPL-UHFFFAOYSA-N
Synonym 6-chloronicotinamide, 3-pyridinecarboxamide, 6-chloro, nicotinamide, 6-chloro, 6-chloro-3-pyridinecarboxamide, 6-chloro-nicotinamide, acmc-1b4gd, 6-chloronicotinic acid amide, 3-carbamoyl-6-chloropyridine, ksc492o5t
PubChem CID 80456
IUPAC Name 6-chloropyridine-3-carboxamide
SMILES NC(=O)C1=CC=C(Cl)N=C1

Specifications

Melting Point 214°C to 216°C
Quantity 1 g
Beilstein 116043
Formula Weight 156.57
Percent Purity 98%
Chemical Name or Material 6-Chloronicotinamide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.