missing translation for 'onlineSavingsMsg'
Learn More
Learn More
6-Chloroindolin-2-one, Mikromol™
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
Description
6-Chloroindolin-2-one
Specifications
Specifications
| Chemical Name or Material | 6-Chloroindolin-2-one |
| CAS | 56341-37-8 |
| Analyte or Component Names | 6-Chloroindolin-2-one |
| Shipping Condition | Room Temperature |
| Molecular Formula | C8 H6 Cl N O |
| Certifications/Compliance | ISO 9001 |
| Synonym | 2H-Indol-2-one, 6-chloro-1,3-dihydro-, 6-Chloro-1,3-dihydro-2H-indol-2-one, 6-Chloro-1,3-dihydroindol-2-one, 6-Chloro-2-indolinone, 6-Chloro-2-oxindole, 6-Chloro-2-oxoindoline, 6-Chlorooxindole |
| InChI Formula | InChI=1S/C8H6ClNO/c9-6-2-1-5-3-8(11)10-7(5)4-6/h1-2,4H,3H2,(H,10,11) |
| SMILES | Clc1ccc2CC(=O)Nc2c1 |
| IUPAC Name | 6-chloroindolin-2-one |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction