missing translation for 'onlineSavingsMsg'
Learn More

6-Chloroimidazo[2,1-b]thiazole-5-sulfonyl chloride, 97%, Thermo Scientific Chemicals

Product Code. 15438585
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15438585 5 g 5g
15428585 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15438585 Supplier Thermo Scientific Alfa Aesar Supplier No. H33683.06

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

It is used as pharmaceutical intermediate. Benzoylthiophenes are allosteric enhancers (AE) of agonist activity at the A1 adenosine receptor.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as pharmaceutical intermediate. Benzoylthiophenes are allosteric enhancers (AE) of agonist activity at the A1 adenosine receptor.

Solubility
Sparingly soluble in water.(0.26 g/L) (25°C),

Notes
Store in cool dry place. Ensure proper ventilation. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 150020-64-7
Molecular Formula C5H2Cl2N2O2S2
Molecular Weight (g/mol) 257.103
MDL Number MFCD00205139
InChI Key APRATEMYDZDYJD-UHFFFAOYSA-N
Synonym 6-chloroimidazo 2,1-b 1,3 thiazole-5-sulfonyl chloride, 6-chloroimidazo 2,1-b thiazole-5-sulfonyl chloride, 6-chloroimidazo 2,1-b thiazole-5-sulfonylchloride, 6-chloro-imidazo 2,1-b thiazole-5-sulfonyl chloride, imidazo 2,1-b thiazole-5-sulfonylchloride, 6-chloro, acmc-1c5af, 6-chloroimidazo 2,1-b 1,3 thiazole-5-sulfonylchloride, 6-chloro-5-imidazo 2,1-b thiazolesulfonyl chloride, 6-chloro-imidazo 2, 1-b thiazole-5-sulfonyl chloride, imidazo 2,1-b thiazole-5-sulfonylchloride,6-chloro
PubChem CID 2799717
IUPAC Name 6-chloroimidazo[2,1-b][1,3]thiazole-5-sulfonyl chloride
SMILES C1=CSC2=NC(=C(N21)S(=O)(=O)Cl)Cl

Specifications

Melting Point 138°C to 140°C
Quantity 5 g
UN Number UN3261
Sensitivity Moisture sensitive
Solubility Information Sparingly soluble in water.(0.26g/L) (25°C),
Formula Weight 257.12
Percent Purity 97%
Chemical Name or Material 6-Chloroimidazo[2,1-b]thiazole-5-sulfonyl chloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.