missing translation for 'onlineSavingsMsg'
Learn More

6-Chloroimidazo[2,1-b][1,3]thiazole-5-sulfonyl chloride, 97%, Thermo Scientific™

Product Code. 10447062
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
5g
This item is not returnable. View return policy

Product Code. 10447062

Brand: Thermo Scientific Maybridge SPB05853DA

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 150020-64-7
Molecular Formula C5H2Cl2N2O2S2
Molecular Weight (g/mol) 257.103
InChI Key APRATEMYDZDYJD-UHFFFAOYSA-N
PubChem CID 2799717
IUPAC Name 6-chloroimidazo[2,1-b][1,3]thiazole-5-sulfonyl chloride
SMILES C1=CSC2=NC(=C(N21)S(=O)(=O)Cl)Cl

Specifications

Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Synonym 6-chloroimidazo 2,1-b 1,3 thiazole-5-sulfonyl chloride, 6-chloroimidazo 2,1-b thiazole-5-sulfonyl chloride, 6-chloroimidazo 2,1-b thiazole-5-sulfonylchloride, 6-chloro-imidazo 2,1-b thiazole-5-sulfonyl chloride, imidazo 2,1-b thiazole-5-sulfonylchloride, 6-chloro, acmc-1c5af, 6-chloroimidazo 2,1-b 1,3 thiazole-5-sulfonylchloride, 6-chloro-5-imidazo 2,1-b thiazolesulfonyl chloride, 6-chloro-imidazo 2, 1-b thiazole-5-sulfonyl chloride, imidazo 2,1-b thiazole-5-sulfonylchloride,6-chloro
Formula Weight 257.12
Percent Purity 97%
Chemical Name or Material 6-Chloroimidazo[2, 1-b][1, 3]thiazole-5-sulfonyl chloride
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.