missing translation for 'onlineSavingsMsg'
Learn More

6-Chlorohexanoyl chloride, 95%

Product Code. 11357566
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11357566 5 g 5g
11347566 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11357566 Supplier Thermo Scientific Alfa Aesar Supplier No. L11111.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 19347-73-0 | C6H10Cl2O | 169.045 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 19347-73-0
Molecular Formula C6H10Cl2O
Molecular Weight (g/mol) 169.045
MDL Number MFCD00041436
InChI Key WZILXAPNPKMOSA-UHFFFAOYSA-N
Synonym hexanoyl chloride, 6-chloro, 6-chloro-caproyl chloride, acmc-1bt4p, 6-chloro-hexanoyl chloride, 6-chlorocaproic acid chloride, 6-chlorohexanoyl chloride
PubChem CID 88014
IUPAC Name 6-chlorohexanoyl chloride
SMILES C(CCC(=O)Cl)CCCl

Specifications

Density 1.4
Boiling Point 90°C (7 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.4652
Quantity 5 g
UN Number UN3265
Beilstein 1852393
Sensitivity Moisture sensitive
Formula Weight 169.05
Percent Purity 95%
Chemical Name or Material 6-Chlorohexanoyl chloride
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.