missing translation for 'onlineSavingsMsg'
Learn More

6-Chlorobenzimidazole-2-carboxylic acid, 97%, Thermo Scientific Chemicals

Product Code. p-7039410
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15494878 1 g 1g
15404888 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15494878 Supplier Thermo Scientific Alfa Aesar Supplier No. H58648.03
Add to basket
This item is not returnable. View return policy

It is employed as a intermediate for pharmaceutical.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed as a intermediate for pharmaceutical.

Solubility
Slightly soluble in water.

Notes
This products exists as two tautomers as there is a rapid exchange of the proton between the two heterocyclic nitrogens. This renders the 5- and 6- positions on the ring chemically equivalent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 39811-14-8
Molecular Formula C8H5ClN2O2
Molecular Weight (g/mol) 196.59
MDL Number MFCD06739053
InChI Key NZIHMSYSZRFUQJ-UHFFFAOYSA-N
Synonym 6-chloro-1h-benzoimidazole-2-carboxylic acid, 6-chloro-1h-benzo d imidazole-2-carboxylic acid, 5-chloro-1h-benzo d imidazole-2-carboxylic acid, 5-chloro-1h-benzimidazole-2-carboxylic acid, 6-chlorobenzimidazole-2-carboxylic acid, 5-chloro-1h-benzoimidazole-2-carboxylic acid, 1h-benzimidazole-2-carboxylic acid, 6-chloro, 5-chloro-1h-1,3-benzodiazole-2-carboxylic acid, 5-chloro-3h-1,3-benzodiazole-2-carboxylic acid, 6-chloro-1h-1,3-benzodiazole-2-carboxylic acid
PubChem CID 10512110
IUPAC Name 6-chloro-1H-benzimidazole-2-carboxylic acid
SMILES C1=CC2=C(C=C1Cl)NC(=N2)C(=O)O

Specifications

Quantity 1 g
Solubility Information Slightly soluble in water.
Formula Weight 196.59
Percent Purity 97%
Chemical Name or Material 6-Chlorobenzimidazole-2-carboxylic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.