missing translation for 'onlineSavingsMsg'
Learn More

6-Chloro-7-methylchromone, 98%, Thermo Scientific™

Catalog Number 11441640
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
67029-84-9
C10H7ClO2
194.614
MFCD00239401
UQXYHMICNLSDMN-UHFFFAOYSA-N
6-chloro-7-methylchromone, 6-chloro-7-methyl-4h-chromen-4-one, 6-chloro-7-methyl-4h-1-benzopyran-4-one, 4h-1-benzopyran-4-one,6-chloro-7-methyl
688707
6-chloro-7-methylchromen-4-one
CC1=C(C=C2C(=O)C=COC2=C1)Cl
This item is not returnable. View return policy
67029-84-9
C10H7ClO2
194.614
MFCD00239401
UQXYHMICNLSDMN-UHFFFAOYSA-N
6-chloro-7-methylchromone, 6-chloro-7-methyl-4h-chromen-4-one, 6-chloro-7-methyl-4h-1-benzopyran-4-one, 4h-1-benzopyran-4-one,6-chloro-7-methyl
688707
6-chloro-7-methylchromen-4-one
CC1=C(C=C2C(=O)C=COC2=C1)Cl

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point 169°C to 172°C
Quantity 1 g
Formula Weight 194.62
Percent Purity 98%
Chemical Name or Material 6-Chloro-7-methylchromone
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'