missing translation for 'onlineSavingsMsg'
Learn More

6-Chloro-7-iodo-7-deazapurine, 96%

Product Code. 15468007
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15468007 5 g 5g
15458007 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15468007 Supplier Thermo Scientific Alfa Aesar Supplier No. H51128.06
 Restricted Item
Estimated Shipment: 21-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 123148-78-7 | C6H3ClIN3 | 279.465 g/mol

6-Chloro-7-iodo-7-deazapurine is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
6-Chloro-7-iodo-7-deazapurine is used as pharmaceutical intermediate.

Solubility
Insoluble in water.

Notes
Light sensitive. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 123148-78-7
Molecular Formula C6H3ClIN3
Molecular Weight (g/mol) 279.465
MDL Number MFCD09263258
InChI Key CBWBJFJMNBPWAL-UHFFFAOYSA-N
Synonym 4-chloro-5-iodo-7h-pyrrolo 2,3-d pyrimidine, 6-chloro-7-iodo-7-deazapurine, 4-chloro-5-iodo-7h-pyrrol 2,3-d pyrimidine, 4-chloro-5-iodo-1h-pyrrolo 2,3-d pyrimidine, 4-chloro-5-iodopyrrolo 2,3-d pyrimidine, 7h-pyrrolo 2,3-d pyrimidine, 4-chloro-5-iodo, acmc-20agy2, amtns039, 6-chloro-7-iod-7-deazapurine
PubChem CID 14809281
IUPAC Name 4-chloro-5-iodo-7H-pyrrolo[2,3-d]pyrimidine
SMILES C1=C(C2=C(N1)N=CN=C2Cl)I

Specifications

Melting Point ∽180°C (decomposition)
Quantity 5 g
Sensitivity Light sensitive
Solubility Information Insoluble in water.
Formula Weight 279.47
Percent Purity 96%
Chemical Name or Material 6-Chloro-7-iodo-7-deazapurine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.