missing translation for 'onlineSavingsMsg'
Learn More

6-Chloro-7-deazapurine, 98%

Product Code. 11339929
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11339929 5 g 5g
11329929 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11339929 Supplier Thermo Scientific Alfa Aesar Supplier No. B25713.06
 Restricted Item
Estimated Shipment: 23-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 3680-69-1 | C6H4ClN3 | 153.569 g/mol

6-Chloro-7-deazapurine is used for making tofacitinib.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
6-Chloro-7-deazapurine is used for making tofacitinib.

Solubility
Soluble in DMSO, ethyl acetate and methanol.

Notes
Store in cool, dry place in a well sealed container. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3680-69-1
Molecular Formula C6H4ClN3
Molecular Weight (g/mol) 153.569
MDL Number MFCD01686865
InChI Key BPTCCCTWWAUJRK-UHFFFAOYSA-N
Synonym 4-chloro-7h-pyrrolo 2,3-d pyrimidine, 4-chloropyrrolo 2,3-d pyrimidine, 6-chloro-7-deazapurine, 4-chloro-1h-pyrrolo 2,3-d pyrimidine, 7h-pyrrolo 2,3-d pyrimidine, 4-chloro, 1h-pyrrolo 2,3-d pyrimidine, 4-chloro, 6-chloro-7-deazapurine 4-chloro-1h-pyrrolo 2,3-d pyrimidine, 4-chloro-1h-pyrrol 2,3-d pyrimidine
PubChem CID 5356682
IUPAC Name 4-chloro-7H-pyrrolo[2,3-d]pyrimidine
SMILES C1=CNC2=C1C(=NC=N2)Cl

Specifications

Melting Point 190°C to 192°C
Quantity 5 g
Solubility Information Soluble in DMSO,ethyl acetate and methanol.
Formula Weight 153.52
Percent Purity 98%
Chemical Name or Material 6-Chloro-7-deazapurine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.