missing translation for 'onlineSavingsMsg'
Learn More

6-Chloro-3-(phenylsulfonyl)-4-(trifluoromethyl)-2(1H)-quinolinone, TRC

Product Code. 30354411
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Quantity:
10 mg
missing translation for 'unitSize':
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity missing translation for 'unitSize'
30354411 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 missing translation for 'options'
This item is not returnable. View return policy
Product Code. 30354411 missing translation for 'mfr' Toronto Research Chemicals missing translation for 'supplierNo' TRCC36824010MG
 Restricted Item
Estimated Shipment: 12-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 6-Chloro-3-(phenylsulfonyl)-4-(trifluoromethyl)-2(1H)-quinolinone
CAS 253663-85-3
Molecular Formula C16H9ClF3NO3S
Quantity 10 mg
Synonym 6-Chloro-3-(phenylsulfonyl)-4-(trifluoromethyl)-2(1H)-quinolinone (ACI)
InChI Formula InChI=1S/C16H9ClF3NO3S/c17-9-6-7-12-11(8-9)13(16(18,19)20)14(15(22)21-12)25(23,24)10-4-2-1-3-5-10/h1-8H,(H,21,22)
SMILES O=C1NC=2C=CC(Cl)=CC2C(=C1S(=O)(=O)C=3C=CC=CC3)C(F)(F)F
IUPAC Name 6-chloro-3-(phenylsulfonyl)-4-(trifluoromethyl)quinolin-2(1H)-one
Molecular Weight (g/mol) 387.76
Formula Weight 386.99
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.