missing translation for 'onlineSavingsMsg'
Learn More

6-chloro-2H-chromene-3-carboxylic acid, 97%, Thermo Scientific™

Product Code. 10344292
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
Unit Size:
10g
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10344292 10 g 10g
10563212 1 g 1g
10168504 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10344292 Supplier Thermo Scientific Maybridge Supplier No. RDR03723EA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 83823-06-7
Molecular Formula C10H7ClO3
Molecular Weight (g/mol) 210.613
MDL Number MFCD00052362
InChI Key ZRCGKWSNRRTAJY-UHFFFAOYSA-N
Synonym 6-chloro-2h-1-benzopyran-3-carboxylic acid, 2h-1-benzopyran-3-carboxylicacid, 6-chloro, 2h-1-benzopyran-3-carboxylic acid, 6-chloro, maybridge1_005975, acmc-20alzg, chlorochromenecarboxylicacid, #, 6-chloro 2h-benzopyran-3-carboxylic acid, 6-chloro 2h-1-benzopyran-3-carboxylic acid, 6-chloro-2h-1-benzopyran-3-carboxylic
PubChem CID 596929
IUPAC Name 6-chloro-2H-chromene-3-carboxylic acid
SMILES C1C(=CC2=C(O1)C=CC(=C2)Cl)C(=O)O

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber glass bottle
Quantity 10 g
Formula Weight 210.62
Percent Purity 97%
Chemical Name or Material 6-chloro-2H-chromene-3-carboxylic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.