missing translation for 'onlineSavingsMsg'
Learn More

6-Chloro-2-hexanone, 98%

Product Code. 15490298
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15490298 5 g 5g
15400308 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15490298 Supplier Thermo Scientific Alfa Aesar Supplier No. H53462.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 10226-30-9 | C6H11ClO | 134.603 g/mol

6-Chloro-2-hexanone in the synthesis of the keto analogs began with 6-chloro-2-hexanone. Substituted 7-(oxoalkyl)theophyllines were synthesized by alkylation of the respective theophyllines with 6-chloro-2-hexanone.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
6-Chloro-2-hexanone in the synthesis of the keto analogs began with 6-chloro-2-hexanone. Substituted 7-(oxoalkyl)theophyllines were synthesized by alkylation of the respective theophyllines with 6-chloro-2-hexanone.

Solubility
Insoluble in water, soluble in chloroform (chloroform, carbon), n-heptane, ethanol, butanol, etc.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10226-30-9
Molecular Formula C6H11ClO
Molecular Weight (g/mol) 134.603
MDL Number MFCD00191625
InChI Key CMDIDTNMHQUVPE-UHFFFAOYSA-N
Synonym 6-chloro-2-hexanone, 1-chloro-5-hexanone, 2-hexanone, 6-chloro, 1-chlorohexan-5-one, 1-chlorohexane-5-one, 4-chlorobutyl methyl ketone, chlorohexanone, 1-chloro-hexan-5-one, 6-chloro-hexan-2-one, 6-chloranylhexan-2-one
PubChem CID 82468
IUPAC Name 6-chlorohexan-2-one
SMILES CC(=O)CCCCCl

Specifications

Density 1.02
Boiling Point 86°C (16 mmHg)
Flash Point 91°C (196°F)
Refractive Index 1.4435
Quantity 5 g
Solubility Information Insoluble in water,soluble in chloroform (chloroform,carbon),n-heptane,ethanol,butanol,etc.
Formula Weight 134.61
Percent Purity 98%
Chemical Name or Material 6-Chloro-2-hexanone

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.