missing translation for 'onlineSavingsMsg'
Learn More

6-Chloro-2-hexanone, 98%, ACROS Organics™

Product Code. 10245811 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10245811 25 g 25g
10576631 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10245811 Supplier Thermo Scientific Acros Supplier No. 343690250
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 10226-30-9
Molecular Formula C6H11ClO
Molecular Weight (g/mol) 134.603
MDL Number MFCD00191625
InChI Key CMDIDTNMHQUVPE-UHFFFAOYSA-N
Synonym 6-chloro-2-hexanone, 1-chloro-5-hexanone, 2-hexanone, 6-chloro, 1-chlorohexan-5-one, 1-chlorohexane-5-one, 4-chlorobutyl methyl ketone, chlorohexanone, 1-chloro-hexan-5-one, 6-chloro-hexan-2-one, 6-chloranylhexan-2-one
PubChem CID 82468
IUPAC Name 6-chlorohexan-2-one
SMILES CC(=O)CCCCCl

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color Yellow to Brown
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula H3CCO(CH2)4Cl
Refractive Index 1.4425 to 1.4445
Quantity 25 g
Formula Weight 134.61
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 6-Chloro-2-hexanone, 98%
Show More Show Less
compliance-icons
Product Identifier
  • 6-Chloro-2-hexanone
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.