missing translation for 'onlineSavingsMsg'
Learn More

6-Chloro-2,3-difluorobenzoyl chloride, 97%, Thermo Scientific™

Catalog Number 15416767
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
1208078-25-4
C7H2Cl2F2O
210.989
MFCD12922578
CHLDUAMDKJNJKC-UHFFFAOYSA-N
50998364
6-chloro-2,3-difluorobenzoyl chloride
C1=CC(=C(C(=C1F)F)C(=O)Cl)Cl
This item is not returnable. View return policy
1208078-25-4
C7H2Cl2F2O
210.989
MFCD12922578
CHLDUAMDKJNJKC-UHFFFAOYSA-N
50998364
6-chloro-2,3-difluorobenzoyl chloride
C1=CC(=C(C(=C1F)F)C(=O)Cl)Cl

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Quantity 1 g
UN Number UN3265
Sensitivity Moisture sensitive
Formula Weight 211
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 6-Chloro-2,3-difluorobenzoyl chloride
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'