missing translation for 'onlineSavingsMsg'
Learn More

6-Chloro-2,3-difluorobenzaldehyde, 97%, Thermo Scientific™

Catalog Number 15487217
Click to view available options
Quantity:
1 g
This item is not returnable. View return policy
797791-33-4
C7H3ClF2O
176.547
MFCD07368118
YVKZXQCYLHFHIR-UHFFFAOYSA-N
9812909
6-chloro-2,3-difluorobenzaldehyde
C1=CC(=C(C(=C1F)F)C=O)Cl
This item is not returnable. View return policy
797791-33-4
C7H3ClF2O
176.547
MFCD07368118
YVKZXQCYLHFHIR-UHFFFAOYSA-N
9812909
6-chloro-2,3-difluorobenzaldehyde
C1=CC(=C(C(=C1F)F)C=O)Cl

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Color White to Yellow
Quantity 1 g
Sensitivity Air sensitive
Synonym benzaldehyde, 6-chloro-2,3-difluoro, benzaldehyde,6-chloro-2,3-difluoro
Formula Weight 176.55
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 6-Chloro-2,3-difluorobenzaldehyde
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'