missing translation for 'onlineSavingsMsg'
Learn More
Learn More
6-Chloro-1-hydroxy-1H-benzotriazole, 97%, Thermo Scientific™
Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 g
100 g
Unit Size:
100g
5g
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Specifications
| Melting Point | 190.0°C to 195.0°C |
| Assay Percent Range | 96% min. (HPLC) |
| Packaging | Glass bottle |
| Quantity | 100 g |
| Formula Weight | 169.57 |
| Percent Purity | 97% |
| Chemical Name or Material | 6-Chloro-1-hydroxy-1H-benzotriazole |
Safety and Handling
Product Identifier
- 6-Chloro-1-hydroxy-1H-benzotriazole
- Warning
- Acute toxicity Category 4
- H302-Harmful if swallowed.
- P264-Wash thoroughly after handling.
- P270-Do not eat, drink or smoke when using this product.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction