Learn More
Thermo Scientific Acros 6-Chloro-1-hydroxy-1H-benzotriazole; 97%; Acros Organics; 25g; Glass bottle; F.W.: 169.57; C6H4ClN3O; MP: 190.0deg.C to 195.0deg.C; CAS: 26198-19-6
Brand:  Thermo Scientific Acros 448920250
6-Chloro-1-hydroxy-1H-benzotriazole; 97%; Acros Organics; 25g; Glass bottle; F.W.: 169.57; C6H4ClN3O; MP: 190.0deg.C to 195.0deg.C; CAS: 26198-19-6
Code : NEW
Additional Details : CAS Number : 26198-19-6 Weight : 0.03000kg
Product Code. 15197422