missing translation for 'onlineSavingsMsg'
Learn More

6-Chloro-1-hexyne, 98%

Product Code. 15400338
Change view
Click to view available options
Quantity:
5 mL
25 mL
Unit Size:
25mL
5mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15400338 5 mL 5mL
15410338 25 mL 25mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15400338 Supplier Thermo Scientific Alfa Aesar Supplier No. H53476.AA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 10297-06-0 | C6H9Cl | 116.59 g/mol

6-Chloro-1-hexyne is used as an organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
6-Chloro-1-hexyne is used as an organic chemical synthesis intermediate.

Notes
Stable under recommended storage conditions. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10297-06-0
Molecular Formula C6H9Cl
Molecular Weight (g/mol) 116.59
MDL Number MFCD00013697
InChI Key ZUKOCGMVJUXIJA-UHFFFAOYSA-N
Synonym 6-chloro-1-hexyne, 5-hexynechloride, 1-hexyne,6-chloro, 1-hexyne, 6-chloro, 5-hexynyl chloride, 1-chloro-5-hexyne, 6-chloro-hex-1-yne, acmc-1boe0, zukocgmvjuxija-uhfffaoysa
PubChem CID 272001
IUPAC Name 6-chlorohex-1-yne
SMILES ClCCCCC#C

Specifications

Density 0.962
Boiling Point 83°C (95 mmHg)
Flash Point 37°C (99°F)
Linear Formula Cl(CH2)4C≡CH
Refractive Index 1.451
Quantity 5 mL
UN Number UN1993
Formula Weight 116.59
Percent Purity 98%
Chemical Name or Material 6-Chloro-1-hexyne
compliance-icons compliance-icons
Product Identifier
  • 6-Chloro-1-hexyne
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contain: 6-Chloro-1-hexyne

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.