missing translation for 'onlineSavingsMsg'
Learn More

6-chloro-1-hexene 97%, Thermo Scientific™

Catalog Number p-3212268
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
928-89-2
MFCD00039391
BLMIXWDJHNJWDT-UHFFFAOYSA-N
6-chloro-1-hexene, 1-hexene, 6-chloro, 5-hexenyl chloride, 6-chlorohexene, chloro-1-hex, 6-chloro-1-hexen, 1-chloro-5-hexene, 1-chlorohex-5-ene, acmc-20alz4, 5-hexen-1-yl chloride
70233
6-chlorohex-1-ene
C=CCCCCCl
This item is not returnable. View return policy
928-89-2
MFCD00039391
BLMIXWDJHNJWDT-UHFFFAOYSA-N
6-chloro-1-hexene, 1-hexene, 6-chloro, 5-hexenyl chloride, 6-chlorohexene, chloro-1-hex, 6-chloro-1-hexen, 1-chloro-5-hexene, 1-chlorohex-5-ene, acmc-20alz4, 5-hexen-1-yl chloride
70233
6-chlorohex-1-ene
C=CCCCCCl
CAS Min % 96.0
CAS Max % 100.0
Color Colorless to Yellow
Density 0.9070g/mL
Boiling Point 131°C
Flash Point 29°C
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Molecular Formula C6H11Cl
Linear Formula CH2CH(CH2)4Cl
Refractive Index 1.4360 to 1.438
Quantity 5 g
Specific Gravity 0.907
Molecular Weight (g/mol) 118.61
Formula Weight 118.61
Percent Purity ≥96.0%
Physical Form Liquid
Chemical Name or Material 6-Chloro-1-hexene
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 6-Chloro-1-hexene
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'