missing translation for 'onlineSavingsMsg'
Learn More

6-Carbethoxy-2,2,6-trimethylcyclohexanone, 97%, Thermo Scientific™

Product Code. 10217700 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
Packaging:
Glass bottle
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10217700 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10217700 Supplier Thermo Scientific Acros Supplier No. 291890010

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 7507-68-8
CAS Min % 96.0
CAS Max % 100.0
Color Colorless
Boiling Point 55°C to 56°C (3.0 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Molecular Formula C12H20O3
Refractive Index 1.4540 to 1.456
MDL Number MFCD00143496
Quantity 1 g
Synonym 6-carbethoxy-2,2,6-trimethylcyclohexanone, ethyl 1,3,3-trimethyl-2-oxocyclohexanecarboxylate
InChI Key JXCZRFBBEUNPTN-UHFFFAOYSA-N
SMILES CCOC(=O)C1(CCCC(C1=O)(C)C)C
IUPAC Name ethyl 1,3,3-trimethyl-2-oxocyclohexane-1-carboxylate
Molecular Weight (g/mol) 212.29
PubChem CID 344633
Formula Weight 212.29
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 6-Carbethoxy-2, 2, 6-trimethylcyclohexanone, 97%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.