missing translation for 'onlineSavingsMsg'
Learn More

6-Bromoveratraldehyde, 97%

Product Code. 11490124
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11490124 25 g 25g
11480124 5 g 5g
11400134 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11490124 Supplier Thermo Scientific Alfa Aesar Supplier No. A13591.14
 Restricted Item
Estimated Shipment: 25-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 5392-10-9 | C9H9BrO3 | 245.07 g/mol

6-Bromoveratraldehyde was used in the preparation of benzylchromenylamines.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
6-Bromoveratraldehyde was used in the preparation of benzylchromenylamines.

Solubility
Insoluble in water.

Notes
Air Sensitive. Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5392-10-9
Molecular Formula C9H9BrO3
Molecular Weight (g/mol) 245.07
MDL Number MFCD00003301
InChI Key UQQROBHFUDBOOK-UHFFFAOYSA-N

Specifications

Melting Point 147°C to 152°C
Quantity 25 g
Beilstein 2052355
Sensitivity Air Sensitive
Synonym 6-bromoveratraldehyde, 6-bromo-3,4-dimethoxybenzaldehyde, benzaldehyde, 2-bromo-4,5-dimethoxy, 6-bromo veratraldehyde, 2-bromo-4,5-dimethoxybenzalehyde, 4,5-dimethoxy-2-bromobenzaldehyde, 6-bromoveratral, pubchem8209, veratraldehyde, 6-bromo, acmc-2097hh
Solubility Information Insoluble in water.
SMILES COC1=C(C=C(C(=C1)C=O)Br)OC
IUPAC Name 2-bromo-4,5-dimethoxybenzaldehyde
PubChem CID 79351
Formula Weight 245.08
Percent Purity 97%
Chemical Name or Material 6-Bromoveratraldehyde
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.