Learn More
6-Bromoquinoxaline, 97%, Maybridge™
Brand: Maybridge CC11710DA
Code : 31
Additional Details : CAS Number : 50998-17-9 Weight : 0.05100kg
Packaging | Amber Glass Bottle |
---|---|
Quantity | 1g |
Chemical Identifiers
50998-17-9 | |
209.046 | |
NOYFLUFQGFNMRB-UHFFFAOYSA-N | |
610939 | |
C1=CC2=NC=CN=C2C=C1Br |
C8H5BrN2 | |
MFCD00837757 | |
6-bromo-quinoxaline, quinoxaline, 6-bromo, 6-bromobenzopyrazine, acmc-209krf, 6-bromoquinoxaline, ksc269e9f | |
6-bromoquinoxaline |
Specifications
6-Bromoquinoxaline | |
1g | |
96.5 | |
97% | |
MFCD00837757 | |
NOYFLUFQGFNMRB-UHFFFAOYSA-N | |
6-bromoquinoxaline | |
610939 | |
97% |
Amber Glass Bottle | |
50998-17-9 | |
100.0 | |
C8H5BrN2 | |
6-bromo-quinoxaline, quinoxaline, 6-bromo, 6-bromobenzopyrazine, acmc-209krf, 6-bromoquinoxaline, ksc269e9f | |
C1=CC2=NC=CN=C2C=C1Br | |
209.046 | |
209.05 |