missing translation for 'onlineSavingsMsg'
Learn More

6-Bromoquinoline, 97%

Product Code. 15481258
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15481258 1 g 1g
15401268 250 mg 250mg
15491258 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15481258 Supplier Thermo Scientific Alfa Aesar Supplier No. H55111.03
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 5332-25-2 | C9H6BrN | 208.058 g/mol

6-Bromoquinoline is used as fine chemical and pharmaceutical intermediate, used as the coupling reagent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
6-Bromoquinoline is used as fine chemical and pharmaceutical intermediate, used as the coupling reagent.

Solubility
Soluble in acetone, acetonitrile, dichloromethane, ethyl acetate and THF.

Notes
Keep container tightly closed. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5332-25-2
Molecular Formula C9H6BrN
Molecular Weight (g/mol) 208.058
MDL Number MFCD00024023
InChI Key IFIHYLCUKYCKRH-UHFFFAOYSA-N
Synonym quinoline, 6-bromo, 6-bromo-quinoline, 6-bromo quinoline, 6-bromchinolin, 6-bromchinolin;, 6-br-quinoline, 6-bromooquinoline, acmc-20aits, pubchem2374, 6-bromoquinoline
PubChem CID 79243
IUPAC Name 6-bromoquinoline
SMILES C1=CC2=C(C=CC(=C2)Br)N=C1

Specifications

Melting Point 24°C
Density 1.538
Flash Point >110°C (230°F)
Refractive Index 1.663
Quantity 1 g
Solubility Information Soluble in acetone,acetonitrile,dichloromethane,ethyl acetate and THF.
Formula Weight 208.06
Percent Purity 97%
Chemical Name or Material 6-Bromoquinoline

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.