Learn More
6-Bromoquinoline, 97%
Beskrivning
6-Bromoquinoline is used as fine chemical and pharmaceutical intermediate, used as the coupling reagent.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
6-Bromoquinoline is used as fine chemical and pharmaceutical intermediate, used as the coupling reagent.
Solubility
Soluble in acetone, acetonitrile, dichloromethane, ethyl acetate and THF.
Notes
Keep container tightly closed. Store away from oxidizing agents.
Specifikationer
Specifikationer
| Melting Point | 24°C |
| Density | 1.538 |
| Flash Point | >110°C (230°F) |
| Refractive Index | 1.663 |
| Quantity | 250 mg |
| Solubility Information | Soluble in acetone,acetonitrile,dichloromethane,ethyl acetate and THF. |
| Formula Weight | 208.06 |
| Percent Purity | 97% |
| Chemical Name or Material | 6-Bromoquinoline |
RUO – Research Use Only
Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.