missing translation for 'onlineSavingsMsg'
Learn More

(6-bromopyrid-2-yl)tributylstannane, 95%, Thermo Scientific™

Product Code. 10450754
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10450754 5 g Amber Glass Bottle 5g
10769205 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10450754 Supplier Thermo Scientific Maybridge Supplier No. MO08548DE
 Restricted Item
Request Bulk or Custom Formats
Currently Unavailable: We have no information when this product will be available.
View alternative products
This item is not returnable. View return policy

Chemical Identifiers

CAS 189083-81-6
Molecular Formula C17H30BrNSn
Molecular Weight (g/mol) 447.048
MDL Number MFCD11706981
InChI Key ZRPIKMNUVOBWBE-UHFFFAOYSA-N
Synonym 2-bromo-6-tributylstannyl pyridine, pyridine, 2-bromo-6-tributylstannyl, 6-bromopyrid-2-yl tributylstannane, acmc-20ao22, 2-bromopyridin-6-yl tributylstannane, 2-bromo-6-tributylstannyl-pyridine, 2-bromo-6-tributylstannyl pyridine h-nmr
PubChem CID 24787029
IUPAC Name (6-bromopyridin-2-yl)-tributylstannane
SMILES CCCC[Sn](CCCC)(CCCC)C1=CC=CC(=N1)Br

Specifications

CAS Min % 95.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 5 g
Formula Weight 447.05
Percent Purity 95%
Chemical Name or Material (6-Bromopyrid-2-yl)tributylstannane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.