missing translation for 'onlineSavingsMsg'
Learn More
Learn More
6-Bromopicolinic acid, TRC
Click to view available options
Quantity:
1 g
Unit Size:
1g
Specifications
Specifications
| Chemical Name or Material | 6-Bromopicolinic acid |
| CAS | 21190-87-4 |
| Molecular Formula | C6H4BrNO2 |
| Quantity | 1 g |
| InChI Formula | InChI=1S/C6H4BrNO2/c7-5-3-1-2-4(8-5)6(9)10/h1-3H,(H,9,10) |
| SMILES | OC(=O)c1cccc(Br)n1 |
| IUPAC Name | 6-bromopyridine-2-carboxylic acid |
| Molecular Weight (g/mol) | 202.01 |
| Formula Weight | 200.9425 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction