missing translation for 'onlineSavingsMsg'
Learn More

6-Bromooxindole, 97%

Product Code. 10336684 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
500 mg
Packaging:
Glass bottle
Unit Size:
500mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10336684 500 mg Glass bottle 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10336684 Supplier Thermo Scientific Acros Supplier No. 399435000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 99365-40-9 | C8H6BrNO | 212.05 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 99365-40-9
Melting Point 217.0°C to 221.0°C
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Molecular Formula C8H6BrNO
MDL Number MFCD02179605,MFCD22576660
Quantity 500 mg
Synonym 6-bromooxindole, 6-bromoindolin-2-one, 6-bromo-2-oxindole, 6-bromo-1,3-dihydro-2h-indol-2-one, 6-bromo-2-oxyindole, 6-bromo-1,3-dihydro-indol-2-one, 6-bromo-2-indolinone, 6-bromooxindol, 6-bromo-2,3-dihydro-1h-indol-2-one, 2h-indol-2-one, 6-bromo-1,3-dihydro
InChI Key JARRYVQFBQVOBE-UHFFFAOYSA-N
SMILES BrC1=CC=C2CC(=O)NC2=C1
IUPAC Name 6-bromo-1,3-dihydroindol-2-one
Molecular Weight (g/mol) 212.05
PubChem CID 2773289
Formula Weight 212.04
Percent Purity 97%
Chemical Name or Material 6-Bromooxindole
Show More Show Less
compliance-icons
Product Identifier
  • 6-Bromooxindole
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.