missing translation for 'onlineSavingsMsg'
Learn More

6-Bromonicotinic acid, 96%

Product Code. 10199789 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
10 g
Packaging:
Glass bottle
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10199789 1 g Glass bottle 1g
10802081 10 g Glass bottle 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10199789 Supplier Thermo Scientific Acros Supplier No. 435400010
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 6311-35-9 | C6H4BrNO2 | 202.01 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6311-35-9
Molecular Formula C6H4BrNO2
Molecular Weight (g/mol) 202.01
MDL Number MFCD01927100
InChI Key JDJBRMNTXORYEN-UHFFFAOYSA-N
Synonym 6-bromonicotinic acid, 6-bromo nicotinic acid, 2-bromo-5-pyridinecarboxylic acid, 2-bromopyridine-5-carboxylic acid, 6-bromo-3-pyridine carboxylic acid, 6-bromonicotinicacid, 6-bromo-nicotinic acid, 6-bromo-3-pyridinecarboxylic acid, 2-bromo-5-pyridine carboxylic acid, 3-pyridinecarboxylic acid, 6-bromo
PubChem CID 238932
IUPAC Name 6-bromopyridine-3-carboxylic acid
SMILES OC(=O)C1=CC=C(Br)N=C1

Specifications

CAS Min % 95.0
CAS Max % 100.0
Melting Point 179°C to 181°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Quantity 1 g
Formula Weight 202.01
Percent Purity 96%
Physical Form Crystalline Powder
Chemical Name or Material 6-Bromonicotinic acid
Show More Show Less
compliance-icons
Product Identifier
  • 6-Bromonicotinic Acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.