missing translation for 'onlineSavingsMsg'
Learn More

6-(Bromomethyl)-3,4-dihydro-2H-1,5-benzodioxepine, 97%, Thermo Scientific™

Product Code. 10193494
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10193494 5 g 5g
10151302 250 mg 250mg
10495882 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10193494 Supplier Thermo Scientific Maybridge Supplier No. CC00408DE
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 499770-96-6
Molecular Formula C10H11BrO2
Molecular Weight (g/mol) 243.10
MDL Number MFCD05664399
InChI Key QDRNTKFXSFDJCH-UHFFFAOYSA-N
Synonym 6-bromomethyl-3,4-dihydro-2h-1,5-benzodioxepine, 2h-1,5-benzodioxepin, 6-bromomethyl-3,4-dihydro, 6-bromomethyl-3,4-dihydro-1,5-benzodioxepin, 6-bromomethyl-2h,3h,4h-benzo b 1,4-dioxepin
PubChem CID 2794992
IUPAC Name 6-(bromomethyl)-3,4-dihydro-2H-1,5-benzodioxepine
SMILES BrCC1=C2OCCCOC2=CC=C1

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Quantity 5 g
Formula Weight 243.1
Percent Purity 97%
Chemical Name or Material 6-(Bromomethyl)-3, 4-dihydro-2H-1, 5-benzodioxepine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.