missing translation for 'onlineSavingsMsg'
Learn More

(6-Bromohexyloxy)-tert-butyldimethylsilane, 99%

Product Code. 15451888
Change view
Click to view available options
Quantity:
5 mL
25 mL
Unit Size:
25mL
5mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15451888 25 mL 25mL
15441888 5 mL 5mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15451888 Supplier Thermo Scientific Alfa Aesar Supplier No. H55581.AC
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 129368-70-3 | C12H27BrOSi | 295.336 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 129368-70-3
Molecular Formula C12H27BrOSi
Molecular Weight (g/mol) 295.336
MDL Number MFCD01863708
InChI Key PBKXRKYUUXKNSL-UHFFFAOYSA-N
Synonym 6-bromohexyloxy-tert-butyldimethylsilane, 6-bromohexyl oxy tert-butyl dimethylsilane, silane, 6-bromohexyl oxy 1,1-dimethylethyl dimethyl, acmc-20apme, 1-t-butyldimethylsilyloxy-6-bromohexane, 6-bromohexyloxy-tert-butyidimethylsilane, 6-bromohexyloxy tert-butyldimethylsilane, 6-bromohexyl tert-butyldimethylsilyl ether
PubChem CID 4260352
IUPAC Name 6-bromohexoxy-tert-butyl-dimethylsilane
SMILES CC(C)(C)[Si](C)(C)OCCCCCCBr

Specifications

Boiling Point 276°C
Color Colorless
Physical Form Liquid
Quantity 25 mL
Flash Point 110°C (230°F)
Formula Weight 295.34
Percent Purity 99%
Refractive Index 1.457
Density 1.053 g/mL
Chemical Name or Material (6-Bromohexyloxy)-tert-butyldimethylsilane

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.