missing translation for 'onlineSavingsMsg'
Learn More

6-Bromohexanoyl chloride, 97%

Product Code. 11478913
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
500 g
Unit Size:
100g
25g
500g
5g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11478913 100 g 100g
11314649 5 g 5g
11468913 25 g 25g
11488913 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 11478913 Supplier Thermo Scientific Alfa Aesar Supplier No. A11063.22
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 22809-37-6 | C6H10BrClO | 213.499 g/mol

6-Bromohexanoyl chloride is used as intermediates, organic synthesis, chemical research, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
6-Bromohexanoyl chloride is used as intermediates, organic synthesis, chemical research, agrochemicals and dyestuff.

Solubility
Reacts with water.

Notes
Store in cool, dry conditions in well sealed containers. Keep container tightly closed. Moisture Sensitive
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 22809-37-6
Molecular Formula C6H10BrClO
Molecular Weight (g/mol) 213.499
MDL Number MFCD00000761
InChI Key HBPVGJGBRWIVSX-UHFFFAOYSA-N
Synonym hexanoyl chloride, 6-bromo, 6-bromohexanoic acid chloride, 6-bromohexanoylchloride, 6-bromocaproyl chloride, bromohexanoyl chloride, zlchem 435, acmc-209fyy, 6-bromo-hexanoylchloride, 6-bromo-hexanoyl chloride, 6-bromanylhexanoyl chloride
PubChem CID 89848
IUPAC Name 6-bromohexanoyl chloride
SMILES C(CCC(=O)Cl)CCBr

Specifications

Density 1.397
Boiling Point 119°C to 120°C (9 mmHg)
Flash Point >110°C (230°F)
Odor Acrid
Linear Formula Br(CH2)5COCl
Refractive Index 1.4875
Quantity 100 g
UN Number UN3265
Beilstein 1749739
Sensitivity Moisture sensitive
Solubility Information Reacts with water.
Formula Weight 213.5
Percent Purity 97%
Chemical Name or Material 6-Bromohexanoyl chloride
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.