missing translation for 'onlineSavingsMsg'
Learn More

6-Bromohexanoic acid, 98%

Product Code. 11672699 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
11672699 500 g Glass bottle 500g
10523341 25 g Glass bottle 25g
10543531 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11672699 Supplier Thermo Scientific Acros Supplier No. 165675000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 4224-70-8 | C6H11BrO2 | 195.06 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4224-70-8
Molecular Formula C6H11BrO2
Molecular Weight (g/mol) 195.06
InChI Key NVRVNSHHLPQGCU-UHFFFAOYSA-N
Synonym 6-bromocaproic acid, hexanoic acid, 6-bromo, 6-bromo-hexanoic acid, 6-bromo-n-caproic acid, 6-bromo hexanoic acid, epsilon-bromo caproic acid, 6-bromo hexanoicacid, 6bromohexanoic acid, 6-bromo-hexanicacid, 6-bromo-caproic acid
PubChem CID 20210
ChEBI CHEBI:60700
IUPAC Name 6-bromohexanoic acid
SMILES C(CCC(=O)O)CCBr

Specifications

Melting Point 32.0°C to 35.0°C
Color Orange to White
Boiling Point 165.0°C to 170.0°C (20.0 mmHg)
Flash Point 67°C
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula Br(CH2)5CO2H
Quantity 500 g
Beilstein 02,II,287
Formula Weight 195.06
Percent Purity 98%
Physical Form Low Melting Crystals
Chemical Name or Material 6-Bromohexanoic acid
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.