missing translation for 'onlineSavingsMsg'
Learn More

6-Bromobenzothiazole, 97%

Product Code. 15429597
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15429597 250 mg 250mg
15409597 1 g 1g
15419597 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15429597 Supplier Thermo Scientific Alfa Aesar Supplier No. H52823.MD
 Restricted Item
Estimated Shipment: 12-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 53218-26-1 | C7H4BrNS | 214.08 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 53218-26-1
Molecular Formula C7H4BrNS
Molecular Weight (g/mol) 214.08
MDL Number MFCD04115372
InChI Key YJOUISWKEOXIMC-UHFFFAOYSA-N
Synonym 6-bromobenzothiazole, 6-bromobenzo d thiazole, benzothiazole, 6-bromo, 6-bromo-benzothiazole, 6-bromo-benzo d thiazole, pubchem21861, acmc-209l4t, 6-bromanyl-1,3-benzothiazole, benzothiazole, 6-bromo-9ci
PubChem CID 2795171
IUPAC Name 6-bromo-1,3-benzothiazole
SMILES C1=CC2=C(C=C1Br)SC=N2

Specifications

Melting Point 52°C to 54°C
Quantity 250 mg
Formula Weight 214.08
Percent Purity 97%
Chemical Name or Material 6-Bromobenzothiazole

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.