missing translation for 'onlineSavingsMsg'
Learn More

6-Bromo-4-hydroxyquinoline-3-carboxylic acid, 96%, Thermo Scientific™

Product Code. 15401196
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
It is used as a pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Incompatible with strong oxidizing agents. Store at room temperature.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 98948-95-9
Molecular Formula C10H6BrNO3
Molecular Weight (g/mol) 268.066
MDL Number MFCD01569278
InChI Key GIUZUAUCCUFVKW-UHFFFAOYSA-N
Synonym 6-bromo-4-hydroxyquinoline-3-carboxylic acid, 6-bromo-4-oxo-1,4-dihydroquinoline-3-carboxylic acid, 6-bromo-4-hydroxy-3-quinolinecarboxylic acid, 4-hydroxy-6-bromoquinoline-3-carboxylic acid, 3-quinolinecarboxylic acid, 6-bromo-4-hydroxy, 3-quinolinecarboxylic acid, 6-bromo-1,4-dihydro-4-oxo, 6-bromo-4-hydroxyquinoline-3-carboxylicacid, 6-bromo-4-hydroxy-quinoline-3-carboxylic acid, 6-bromo-1,4-dihydro-4-oxoquinoline-3-carboxylic acid, pubchem17804
PubChem CID 693203
IUPAC Name 6-bromo-4-oxo-1H-quinoline-3-carboxylic acid
SMILES C1=CC2=C(C=C1Br)C(=O)C(=CN2)C(=O)O

Specifications

Quantity 250 mg
Solubility Information Slightly soluble in water.
Formula Weight 268.06
Percent Purity 96%
Chemical Name or Material 6-Bromo-4-hydroxyquinoline-3-carboxylic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'