missing translation for 'onlineSavingsMsg'
Learn More
Learn More
6-Bromo-3-pyridazinamine, 97%, Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Amber Glass Bottle
Specifications
CAS Min % | 97.0 |
Assay Percent Range | 97% |
Packaging | Amber Glass Bottle |
Quantity | 250 mg |
Synonym | 3-amino-6-bromopyridazine, 6-bromo-3-pyridazinamine, 6-bromo-pyridazin-3-ylamine, 2-amino-5-bromopyridazine, 3-pyridazinamine,6-bromo, 3-pyridazinamine, 6-bromo, 6-bromopyridazine-3-ylamine, 6-bromo-3-pyridazineamine, 3-bromo-6-aminopyridazine, acmc-20a0gf |
Formula Weight | 174 |
Percent Purity | 97% |
Chemical Name or Material | 6-Bromo-3-pyridazinamine |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'