missing translation for 'onlineSavingsMsg'
Learn More

6-Bromo-2-pyridinecarbonitrile, 98%

Product Code. 10584598 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
Packaging:
Glass bottle
Unit Size:
1g
Product Code. Quantity Packaging unitSize
10584598 1 g Glass bottle 1g
1 options
This item is not returnable. View return policy

Product Code. 10584598

Brand: Thermo Scientific Acros 429990010

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 122918-25-6 | C6H3BrN2 | 183.01 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 122918-25-6
CAS Min % 97.5
CAS Max % 100.0
Melting Point 98.5°C to 101.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Molecular Formula C6H3BrN2
MDL Number MFCD00234142
Quantity 1 g
Synonym 6-bromopicolinonitrile, 6-bromo-2-pyridinecarbonitrile, 2-bromo-6-cyanopyridine, 6-bromo-2-cyanopyridine, 2-pyridinecarbonitrile, 6-bromo, 2-cyano-6-bromopyridine, 2-bromopyridine-6-carbonitrile, pubchem14367, 2-bromo-6-cyanopyridin, 2-bromo-6-cyano pyridine
InChI Key HNEBPTAKURBYRM-UHFFFAOYSA-N
SMILES C1=CC(=NC(=C1)Br)C#N
IUPAC Name 6-bromopyridine-2-carbonitrile
Molecular Weight (g/mol) 183.01
PubChem CID 15100859
Formula Weight 183.01
Percent Purity 98%
Physical Form Crystalline Powder or Low Melting Solid
Chemical Name or Material 6-Bromo-2-pyridinecarbonitrile
Show More Show Less
compliance-icons
Product Identifier
  • 6-Bromo-2-pyridinecarbonitrile
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.