missing translation for 'onlineSavingsMsg'
Learn More

6-Bromo-2-naphthol, 97%

Product Code. 11488447
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11488447 10 g 10g
11498447 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11488447 Supplier Thermo Scientific Alfa Aesar Supplier No. A18535.09
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 15231-91-1 | C10H7BrO | 223.069 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 15231-91-1
Molecular Formula C10H7BrO
Molecular Weight (g/mol) 223.069
MDL Number MFCD00004081
InChI Key YLDFTMJPQJXGSS-UHFFFAOYSA-N
Synonym 6-bromo-2-naphthol, 2-naphthalenol, 6-bromo, 6-bromo-naphthalen-2-ol, 6-bromo-2-naphthalenol, bromo-6 naphtol-2, 6-bromo2-hydroxynaphthalene, unii-87f10ahz3o, bromo-6 naphtol-2 french, 2-naphthol, 6-bromo, 6-bromo-beta-naphthol
PubChem CID 27144
ChEBI CHEBI:34466
IUPAC Name 6-bromonaphthalen-2-ol
SMILES C1=CC2=C(C=CC(=C2)Br)C=C1O

Specifications

Melting Point 126°C to 129°C
Quantity 10 g
Beilstein 1100270
Formula Weight 223.08
Percent Purity 97%
Chemical Name or Material 6-Bromo-2-naphthol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.