missing translation for 'onlineSavingsMsg'
Learn More

6-Bromo-2-naphthol, 97%

Product Code. 10696324 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10696324 100 g Glass bottle 100g
10335930 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10696324 Supplier Thermo Scientific Acros Supplier No. 107171000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 15231-91-1 | C10H7BrO | 223.07 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 15231-91-1
Molecular Formula C10H7BrO
Molecular Weight (g/mol) 223.07
MDL Number MFCD00004081
InChI Key YLDFTMJPQJXGSS-UHFFFAOYSA-N
Synonym 6-bromo-2-naphthol, 2-naphthalenol, 6-bromo, 6-bromo-naphthalen-2-ol, 6-bromo-2-naphthalenol, bromo-6 naphtol-2, 6-bromo2-hydroxynaphthalene, unii-87f10ahz3o, bromo-6 naphtol-2 french, 2-naphthol, 6-bromo, 6-bromo-beta-naphthol
PubChem CID 27144
ChEBI CHEBI:34466
IUPAC Name 6-bromonaphthalen-2-ol
SMILES C1=CC2=C(C=CC(=C2)Br)C=C1O

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 125°C to 130°C
Color White to Beige
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Linear Formula BrC10H6OH
Quantity 100 g
Beilstein 06, 651
Formula Weight 223.07
Percent Purity 97%
Physical Form Powder
Chemical Name or Material 6-Bromo-2-naphthol
Show More Show Less
compliance-icons
Product Identifier
  • 6-Bromo-2-naphthol
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.