missing translation for 'onlineSavingsMsg'
Learn More

6-Bromo-2-naphthoic acid, 98%

Product Code. 11362887
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11362887 25 g 25g
11352887 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11362887 Supplier Thermo Scientific Alfa Aesar Supplier No. L16093.14
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 5773-80-8 | C11H7BrO2 | 251.079 g/mol

Benzimidazole derivatives has been prepared by using ortho-phenyl diamine and 6-bromo-2-naphthoic acid.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Benzimidazole derivatives has been prepared by using ortho-phenyl diamine and 6-bromo-2-naphthoic acid.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5773-80-8
Molecular Formula C11H7BrO2
Molecular Weight (g/mol) 251.079
MDL Number MFCD01075720
InChI Key NPMCAVBMOTZUPD-UHFFFAOYSA-N
Synonym 6-bromo-2-naphthoic acid, 2-naphthalenecarboxylic acid, 6-bromo, 6-bromo-2-naphtholic aicd, 6-bromo-2-naphthalenecarboxylic acid, 6-bromo-2-naphthalene carboxylic acid, 6-bromanylnaphthalene-2-carboxylic acid, pubchem9466, 6-bromo-2-naphthoicacid, ksc269g5d, 6-bromo-2-naphthylacetic acid
PubChem CID 4549852
IUPAC Name 6-bromonaphthalene-2-carboxylic acid
SMILES C1=CC2=C(C=CC(=C2)Br)C=C1C(=O)O

Specifications

Melting Point 292°C to 296°C
Quantity 25 g
Beilstein 2328940
Solubility Information Insoluble in water.
Formula Weight 251.08
Percent Purity 98%
Chemical Name or Material 6-Bromo-2-naphthoic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.