missing translation for 'onlineSavingsMsg'
Learn More

6-Bromo-2-methylquinoline, 97%, Thermo Scientific Chemicals

Catalog Number 11347299
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
877-42-9
C10H8BrN
222.085
MFCD00079724
SQRYQSKJZVQJAY-UHFFFAOYSA-N
6-bromoquinaldine, 2-methyl-6-bromoquinoline, 6-bromo-2-methyl-quinoline, quinoline, 6-bromo-2-methyl, 6-bromo-2-methyl-1-azanaphthalene, zlchem 926, pubchem5882, acmc-209qps, ksc494e7l
522885
6-bromo-2-methylquinoline
CC1=NC2=C(C=C1)C=C(C=C2)Br
This item is not returnable. View return policy
877-42-9
C10H8BrN
222.085
MFCD00079724
SQRYQSKJZVQJAY-UHFFFAOYSA-N
6-bromoquinaldine, 2-methyl-6-bromoquinoline, 6-bromo-2-methyl-quinoline, quinoline, 6-bromo-2-methyl, 6-bromo-2-methyl-1-azanaphthalene, zlchem 926, pubchem5882, acmc-209qps, ksc494e7l
522885
6-bromo-2-methylquinoline
CC1=NC2=C(C=C1)C=C(C=C2)Br

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 100°C to 103°C
Quantity 5 g
Formula Weight 222.08
Percent Purity 97%
Chemical Name or Material 6-Bromo-2-methylquinoline

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'