missing translation for 'onlineSavingsMsg'
Learn More

6-Bromo-2-methylpyrazolo[1,5-a]pyrimidine, 97%, Thermo Scientific™

Product Code. 15416737
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15416737 1 g 1g
15426737 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15416737 Supplier Thermo Scientific Alfa Aesar Supplier No. H33857.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 916256-65-0
Molecular Formula C7H6BrN3
Molecular Weight (g/mol) 212.05
MDL Number MFCD09863428
InChI Key JUWPPXBKRXAJMY-UHFFFAOYSA-N
PubChem CID 15980041
IUPAC Name 6-bromo-2-methylpyrazolo[1,5-a]pyrimidine
SMILES CC1=NN2C=C(C=NC2=C1)Br

Specifications

Melting Point 137°C to 138°C
Quantity 1 g
Sensitivity Light sensitive
Synonym 6-bromo-2-methylpyrazolo 1,5-a pyrimidine, pyrazolo 1,5-a pyrimidine, 6-bromo-2-methyl, pyrazolo 1,5-a pyrimidine,6-bromo-2-methyl
Formula Weight 212.05
Percent Purity 97%
Chemical Name or Material 6-Bromo-2-methylpyrazolo[1,5-a]pyrimidine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.