missing translation for 'onlineSavingsMsg'
Learn More

6-Bromo-2-methyl-3-nitropyridine, 97%

Product Code. 15431439
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15431439 25 g 25g
15421439 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15431439 Supplier Thermo Scientific Alfa Aesar Supplier No. H64299.14
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 22282-96-8 | C6H5BrN2O2 | 217.02 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 22282-96-8
Molecular Formula C6H5BrN2O2
Molecular Weight (g/mol) 217.02
MDL Number MFCD03095219
InChI Key PCNVKBBKROTUNS-UHFFFAOYSA-N
Synonym 2-bromo-5-nitro-6-picoline, 2-bromo-6-methyl-5-nitropyridine, 2-bromo-5-nitro-6-methylpyridine, 6-bromo-2-methyl-3-nitro-pyridine, 6-bromo-3-nitro-2-picoline, 6-bromo-3-nitropicoline, 6-brom-2-methyl-3-nitro-pyridin, pubchem2968, acmc-209fuh, ksc549a9p
PubChem CID 24728014
IUPAC Name 6-bromo-2-methyl-3-nitropyridine
SMILES CC1=C(C=CC(Br)=N1)[N+]([O-])=O

Specifications

Melting Point 67°C to 72°C
Quantity 25 g
Beilstein 135841
Formula Weight 217.02
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 6-Bromo-2-methyl-3-nitropyridine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.