missing translation for 'onlineSavingsMsg'
Learn More

6-Bromo-2-methyl-3-nitropyridine, 97%

Product Code. 15421439
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 15421439

Brand: Thermo Scientific Alfa Aesar H64299.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 22282-96-8 | C6H5BrN2O2 | 217.02 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 22282-96-8
Molecular Formula C6H5BrN2O2
Molecular Weight (g/mol) 217.02
MDL Number MFCD03095219
InChI Key PCNVKBBKROTUNS-UHFFFAOYSA-N
Synonym 2-bromo-5-nitro-6-picoline, 2-bromo-6-methyl-5-nitropyridine, 2-bromo-5-nitro-6-methylpyridine, 6-bromo-2-methyl-3-nitro-pyridine, 6-bromo-3-nitro-2-picoline, 6-bromo-3-nitropicoline, 6-brom-2-methyl-3-nitro-pyridin, pubchem2968, acmc-209fuh, ksc549a9p
PubChem CID 24728014
IUPAC Name 6-bromo-2-methyl-3-nitropyridine
SMILES CC1=C(C=CC(Br)=N1)[N+]([O-])=O

Specifications

Melting Point 67°C to 72°C
Quantity 5 g
Beilstein 135841
Formula Weight 217.02
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 6-Bromo-2-methyl-3-nitropyridine

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.