missing translation for 'onlineSavingsMsg'
Learn More

6-Bromo-2-chloroquinoline, 96%

Product Code. 15481048
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15481048 5 g 5g
15491048 250 mg 250mg
15471048 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15481048 Supplier Thermo Scientific Alfa Aesar Supplier No. H54895.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1810-71-5 | C9H5BrClN | 242.50 g/mol

It is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as a pharmaceutical intermediate.

Solubility
Soluble in Chloroform and Methanol

Notes
Store at room temperature with proper ventilation.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1810-71-5
Molecular Formula C9H5BrClN
Molecular Weight (g/mol) 242.50
MDL Number MFCD04115272
InChI Key YXRDWUJAJLDABJ-UHFFFAOYSA-N
PubChem CID 12894086
IUPAC Name 6-bromo-2-chloroquinoline
SMILES ClC1=NC2=CC=C(Br)C=C2C=C1

Specifications

Melting Point 148°C to 150°C
Quantity 5 g
Synonym 6-bromo-2-chloro-quinoline, 2-chloro-6-bromoquinoline, quinoline, 6-bromo-2-chloro, pubchem14631, acmc-209ehe, 6-bromo-2 chloroquinoline, 2-chloro-6-bromo-quinoline, ksc537o9r, quinoline,6-bromo-2-chloro, abbypharma ap-14-10831
Solubility Information Soluble in Chloroform and Methanol
Formula Weight 242.5
Percent Purity 96%
Chemical Name or Material 6-Bromo-2-chloroquinoline

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.