missing translation for 'onlineSavingsMsg'
Learn More

6-Bromo-2-benzothiazolinone, 98%, Thermo Scientific™

Product Code. 10525691 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10525691 5 g Glass bottle 5g
10021971 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10525691 Supplier Thermo Scientific Acros Supplier No. 339880050

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 62266-82-4
Molecular Formula C7H4BrNOS
Molecular Weight (g/mol) 230.08
MDL Number MFCD00239363
InChI Key HECJMTPEVWQFCY-UHFFFAOYSA-N
Synonym 6-bromo-2-benzothiazolinone, 6-bromobenzo d thiazol-2 3h-one, 6-bromo-2 3h-benzothiazolone, 6-bromo-3h-benzothiazol-2-one, 2 3h-benzothiazolone, 6-bromo, 6-bromo-1,3-benzothiazol-2 3h-one, 6-bromo-2-hydroxy benzothiazol, 6-bromo-2,3-dihydro-1,3-benzothiazol-2-one, 2-benzothiazolol, 6-bromo, pubchem21839
PubChem CID 188444
SMILES BrC1=CC=C2NC(=O)SC2=C1

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 228°C to 232°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Quantity 5 g
IUPAC Name 6-bromo-2,3-dihydro-1,3-benzothiazol-2-one
Formula Weight 230.08
Percent Purity ≥97.5%
Physical Form Crystalline Powder
Chemical Name or Material 6-Bromo-2-benzothiazolinone, 98%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.